Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRRF
Entrez Gene ID
92399
Gene Name
mitochondrial ribosome recycling factor
Gene Aliases
MRFF, MTRRF, RRF
Chromosome Location
Chr.9: 122264603 - 122323464 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138777.3 NP_620132.1 3 - 4 702 146
NM_199177.2 NP_954646.1 2 - 3 366 146
XM_005252302.3 XP_005252359.1 4 - 5 770 146
XM_011519183.2 XP_011517485.1 4 - 5 1025 146
XM_011519184.2 XP_011517486.1 4 - 5 535 146
XM_011519185.2 XP_011517487.1 4 - 5 770 146
XM_011519186.1 XP_011517488.1 3 - 4 450 146
XM_011519187.2 XP_011517489.1 2 - 3 490 146
XM_017015286.1 XP_016870775.1 4 - 5 770 146
XM_017015287.1 XP_016870776.1 3 - 4 450 146
GenBank mRNA AK291603.1 - 3 - 4 392 146
AK291809.1 - 3 - 4 431 146
AK292790.1 - 3 - 4 431 146
AK298046.1 - 3 - 4 421 146
AK310974.1 - 2 - 3 490 146
AK316266.1 - 4 - 5 719 146
BC013049.1 - 2 - 3 363 146
BC053604.1 - 2 - 3 364 146
BF982227.1 - - 437 146

View Full Product Details