Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRRF
Entrez Gene ID
92399
Gene Name
mitochondrial ribosome recycling factor
Gene Aliases
MRFF, MTRRF, RRF
Chromosome Location
Chr.9: 122264603 - 122323464 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK291809.1 - 4 - 5 547 91
AK292790.1 - 4 - 5 547 91

View Full Product Details