Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRRF
Entrez Gene ID
92399
Gene Name
mitochondrial ribosome recycling factor
Gene Aliases
MRFF, MTRRF, RRF
Chromosome Location
Chr.9: 122264603 - 122323464 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001173512.1 NP_001166983.1 3 - 4 674 97
NM_138777.3 NP_620132.1 4 - 5 830 97
NM_199177.2 NP_954646.1 3 - 4 494 97
XM_005252302.3 XP_005252359.1 5 - 6 898 97
XM_011519183.2 XP_011517485.1 5 - 6 1153 97
XM_011519184.2 XP_011517486.1 5 - 6 663 97
XM_011519185.2 XP_011517487.1 5 - 6 898 97
XM_011519186.1 XP_011517488.1 4 - 5 578 97
XM_011519187.2 XP_011517489.1 3 - 4 618 97
GenBank mRNA AK291603.1 - 4 - 5 520 97
AK300718.1 - 3 - 4 377 97
BC013049.1 - 3 - 4 491 97
BC053604.1 - 3 - 4 492 97
BF982227.1 - - 565 97

View Full Product Details