Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZXDC
Entrez Gene ID
79364
Gene Name
ZXD family zinc finger C
Gene Aliases
ZXDL
Chromosome Location
Chr.3: 126437348 - 126475938 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025112.4 NP_079388.3 6 - 7 2184 61
XM_005247757.3 XP_005247814.1 6 - 7 2195 61
XM_011513119.2 XP_011511421.1 5 - 6 2032 61
GenBank mRNA AK023923.1 - 3 - 4 886 61
AY277596.1 - 2 - 3 746 61
AY936556.1 - 6 - 7 2144 61
BC002940.2 - 5 - 6 1122 61
BC012729.2 - 5 - 6 1122 61
DQ079590.1 - 6 - 7 2184 61

View Full Product Details