The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014510.2 | NP_055325.2 | 17 - 18 | 14630 | 126 |
NM_033026.5 | NP_149015.2 | 17 - 18 | 14630 | 126 | |
XM_017012006.1 | XP_016867495.1 | 15 - 16 | 8810 | 126 | |
GenBank mRNA | AB011131.1 | - | 11 - 12 | 3174 | 126 |