The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282750.1 | NP_001269679.1 | 4 - 5 | 785 | 139 |
NM_001282751.1 | NP_001269680.1 | 5 - 6 | 896 | 139 | |
NM_014283.4 | NP_055098.1 | 5 - 6 | 896 | 139 | |
NM_016227.3 | NP_057311.3 | 5 - 6 | 1176 | 139 | |
XM_006711374.2 | XP_006711437.1 | 5 - 6 | 977 | 139 | |
XM_006711375.2 | XP_006711438.1 | 4 - 5 | 866 | 139 | |
XM_006711376.1 | XP_006711439.1 | 4 - 5 | 866 | 139 | |
GenBank mRNA | AF097535.1 | - | 5 - 6 | 1176 | 139 |
AJ250075.1 | - | 5 - 6 | 894 | 139 | |
AK302509.1 | - | 4 - 5 | 720 | 139 | |
AK302949.1 | - | 5 - 6 | 896 | 139 | |
AL035291.1 | - | 5 - 6 | 797 | 139 | |
BC137125.1 | - | 5 - 6 | 682 | 139 | |
BC144487.1 | - | 5 - 6 | 682 | 139 |