Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHD9
Entrez Gene ID
80205
Gene Name
chromodomain helicase DNA binding protein 9
Gene Aliases
AD013, CHD-9, CReMM, KISH2, PRIC320
Chromosome Location
Chr.16: 53054991 - 53327502 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308319.1 NP_001295248.1 19 - 20 4486 125
NM_025134.4 NP_079410.4 20 - 21 4531 125
XM_005256168.3 XP_005256225.1 20 - 21 4573 125
XM_005256169.2 XP_005256226.1 20 - 21 4595 125
XM_005256172.3 XP_005256229.1 20 - 21 4512 125
XM_005256176.3 XP_005256233.1 19 - 20 3180 125
XM_006721281.2 XP_006721344.1 20 - 21 4512 125
XM_006721283.3 XP_006721346.1 20 - 21 4733 125
XM_011523347.2 XP_011521649.1 21 - 22 4645 125
XM_017023718.1 XP_016879207.1 21 - 22 4768 125
XM_017023719.1 XP_016879208.1 22 - 23 4719 125
XM_017023720.1 XP_016879209.1 21 - 22 4613 125
XM_017023721.1 XP_016879210.1 21 - 22 4769 125
XM_017023722.1 XP_016879211.1 21 - 22 4769 125
XM_017023723.1 XP_016879212.1 21 - 22 4769 125
XM_017023724.1 XP_016879213.1 21 - 22 4595 125
XM_017023725.1 XP_016879214.1 21 - 22 4595 125
XM_017023726.1 XP_016879215.1 19 - 20 3802 125
XM_017023727.1 XP_016879216.1 19 - 20 3180 125
XM_017023728.1 XP_016879217.1 21 - 22 4629 125
GenBank mRNA AB002306.3 - 19 - 20 3954 125
AK299045.1 - 19 - 20 3229 125
AY243500.1 - 19 - 20 4409 125
AY647157.1 - 20 - 21 4531 125
BC140815.1 - 19 - 20 4363 125
BC144622.1 - 19 - 20 4363 125

View Full Product Details