The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308319.1 | NP_001295248.1 | 22 - 23 | 5047 | 151 |
NM_025134.4 | NP_079410.4 | 23 - 24 | 5092 | 151 | |
XM_005256168.3 | XP_005256225.1 | 23 - 24 | 5134 | 151 | |
XM_005256169.2 | XP_005256226.1 | 23 - 24 | 5156 | 151 | |
XM_005256172.3 | XP_005256229.1 | 23 - 24 | 5073 | 151 | |
XM_005256176.3 | XP_005256233.1 | 22 - 23 | 3741 | 151 | |
XM_006721281.2 | XP_006721344.1 | 23 - 24 | 5073 | 151 | |
XM_006721283.3 | XP_006721346.1 | 23 - 24 | 5294 | 151 | |
XM_011523347.2 | XP_011521649.1 | 24 - 25 | 5206 | 151 | |
XM_017023718.1 | XP_016879207.1 | 24 - 25 | 5329 | 151 | |
XM_017023719.1 | XP_016879208.1 | 25 - 26 | 5280 | 151 | |
XM_017023720.1 | XP_016879209.1 | 24 - 25 | 5174 | 151 | |
XM_017023721.1 | XP_016879210.1 | 24 - 25 | 5330 | 151 | |
XM_017023722.1 | XP_016879211.1 | 24 - 25 | 5330 | 151 | |
XM_017023723.1 | XP_016879212.1 | 24 - 25 | 5330 | 151 | |
XM_017023724.1 | XP_016879213.1 | 24 - 25 | 5156 | 151 | |
XM_017023725.1 | XP_016879214.1 | 24 - 25 | 5156 | 151 | |
XM_017023726.1 | XP_016879215.1 | 22 - 23 | 4363 | 151 | |
XM_017023727.1 | XP_016879216.1 | 22 - 23 | 3741 | 151 | |
XM_017023728.1 | XP_016879217.1 | 24 - 25 | 5190 | 151 | |
GenBank mRNA | AB002306.3 | - | 22 - 23 | 4515 | 151 |
AY243500.1 | - | 22 - 23 | 4970 | 151 | |
AY243501.1 | - | 4 - 5 | 652 | 151 | |
AY647157.1 | - | 23 - 24 | 5092 | 151 | |
BC140815.1 | - | 22 - 23 | 4924 | 151 | |
BC144622.1 | - | 22 - 23 | 4924 | 151 | |
DQ059482.1 | - | 13 - 14 | 2420 | 151 | |
DQ333316.1 | - | 23 - 24 | 5186 | 151 |