Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHD9
Entrez Gene ID
80205
Gene Name
chromodomain helicase DNA binding protein 9
Gene Aliases
AD013, CHD-9, CReMM, KISH2, PRIC320
Chromosome Location
Chr.16: 53054991 - 53327502 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308319.1 NP_001295248.1 4 - 5 2205 69
NM_025134.4 NP_079410.4 5 - 6 2250 69
XM_005256168.3 XP_005256225.1 5 - 6 2292 69
XM_005256169.2 XP_005256226.1 5 - 6 2314 69
XM_005256172.3 XP_005256229.1 5 - 6 2231 69
XM_005256176.3 XP_005256233.1 4 - 5 899 69
XM_006721281.2 XP_006721344.1 5 - 6 2231 69
XM_006721283.3 XP_006721346.1 5 - 6 2452 69
XM_011523347.2 XP_011521649.1 6 - 7 2364 69
XM_017023718.1 XP_016879207.1 6 - 7 2487 69
XM_017023719.1 XP_016879208.1 7 - 8 2438 69
XM_017023720.1 XP_016879209.1 6 - 7 2332 69
XM_017023721.1 XP_016879210.1 6 - 7 2488 69
XM_017023722.1 XP_016879211.1 6 - 7 2488 69
XM_017023723.1 XP_016879212.1 6 - 7 2488 69
XM_017023724.1 XP_016879213.1 6 - 7 2314 69
XM_017023725.1 XP_016879214.1 6 - 7 2314 69
XM_017023726.1 XP_016879215.1 4 - 5 1521 69
XM_017023727.1 XP_016879216.1 4 - 5 899 69
XM_017023728.1 XP_016879217.1 6 - 7 2348 69
GenBank mRNA AB002306.3 - 4 - 5 1673 69
AK299045.1 - 4 - 5 948 69
AY243500.1 - 4 - 5 2128 69
BC140815.1 - 4 - 5 2082 69
BC144622.1 - 4 - 5 2082 69
BX390832.2 - - 94 69
DQ333316.1 - 5 - 6 2344 69

View Full Product Details