Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHD9
Entrez Gene ID
80205
Gene Name
chromodomain helicase DNA binding protein 9
Gene Aliases
AD013, CHD-9, CReMM, KISH2, PRIC320
Chromosome Location
Chr.16: 53054991 - 53327502 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308319.1 NP_001295248.1 6 - 7 2336 151
NM_025134.4 NP_079410.4 7 - 8 2381 151
XM_005256168.3 XP_005256225.1 7 - 8 2423 151
XM_005256169.2 XP_005256226.1 7 - 8 2445 151
XM_005256172.3 XP_005256229.1 7 - 8 2362 151
XM_005256176.3 XP_005256233.1 6 - 7 1030 151
XM_006721281.2 XP_006721344.1 7 - 8 2362 151
XM_006721283.3 XP_006721346.1 7 - 8 2583 151
XM_011523347.2 XP_011521649.1 8 - 9 2495 151
XM_017023718.1 XP_016879207.1 8 - 9 2618 151
XM_017023719.1 XP_016879208.1 9 - 10 2569 151
XM_017023720.1 XP_016879209.1 8 - 9 2463 151
XM_017023721.1 XP_016879210.1 8 - 9 2619 151
XM_017023722.1 XP_016879211.1 8 - 9 2619 151
XM_017023723.1 XP_016879212.1 8 - 9 2619 151
XM_017023724.1 XP_016879213.1 8 - 9 2445 151
XM_017023725.1 XP_016879214.1 8 - 9 2445 151
XM_017023726.1 XP_016879215.1 6 - 7 1652 151
XM_017023727.1 XP_016879216.1 6 - 7 1030 151
XM_017023728.1 XP_016879217.1 8 - 9 2479 151
GenBank mRNA AB002306.3 - 6 - 7 1804 151
AK299045.1 - 6 - 7 1079 151
AY243500.1 - 6 - 7 2259 151
BC140815.1 - 6 - 7 2213 151
BC144622.1 - 6 - 7 2213 151
BX390832.2 - - 225 151
DQ333316.1 - 7 - 8 2475 151

View Full Product Details