The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286734.1 | NP_001273663.1 | 31 - 32 | 4865 | 154 |
NM_001813.2 | NP_001804.2 | 32 - 33 | 4940 | 154 | |
XM_011531544.2 | XP_011529846.1 | 31 - 32 | 4858 | 154 | |
XM_011531545.2 | XP_011529847.1 | 32 - 33 | 4939 | 154 | |
XM_011531546.2 | XP_011529848.1 | 31 - 32 | 4735 | 154 | |
XM_011531547.2 | XP_011529849.1 | 32 - 33 | 4939 | 154 | |
XM_011531548.2 | XP_011529850.1 | 32 - 33 | 4939 | 154 | |
XM_017007659.1 | XP_016863148.1 | 32 - 33 | 4939 | 154 | |
GenBank mRNA | AB209996.1 | - | 31 - 32 | 4790 | 154 |
AL596934.1 | - | 2 - 3 | 349 | 154 | |
Z15005.1 | - | 32 - 33 | 4940 | 154 |