The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005956.3 | NP_005947.3 | 2 - 3 | 509 | 97 |
GenBank mRNA | AK302747.1 | - | 2 - 3 | 519 | 97 |
AK312361.1 | - | 2 - 3 | 214 | 97 | |
BC009806.2 | - | 2 - 3 | 175 | 97 | |
BC050420.1 | - | 2 - 3 | 200 | 97 | |
BQ959308.1 | - | 2 - 3 | 321 | 97 | |
J04031.1 | - | 2 - 3 | 175 | 97 |