Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COL19A1
Entrez Gene ID
1310
Gene Name
collagen type XIX alpha 1 chain
Gene Aliases
COL9A1L, D6S228E
Chromosome Location
Chr.6: 69866556 - 70215063 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001858.5 NP_001849.2 28 - 29 2045 87
XM_011535437.2 XP_011533739.1 28 - 29 2312 87
XM_011535438.2 XP_011533740.1 28 - 29 2040 87
XM_017010253.1 XP_016865742.1 27 - 28 2270 87
XM_017010254.1 XP_016865743.1 27 - 28 2258 87
XM_017010255.1 XP_016865744.1 28 - 29 2251 87
XM_017010256.1 XP_016865745.1 27 - 28 2003 87
XM_017010257.1 XP_016865746.1 27 - 28 2186 87
XM_017010258.1 XP_016865747.1 27 - 28 1919 87
XM_017010259.1 XP_016865748.1 28 - 29 2311 87
GenBank mRNA BC113362.1 - 28 - 29 2013 87
BC113364.1 - 28 - 29 2013 87
D38163.1 - 28 - 29 2045 87
L12347.1 - 24 - 25 1613 87

View Full Product Details