Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL19A1
Entrez Gene ID
1310
Gene Name
collagen type XIX alpha 1 chain
Gene Aliases
COL9A1L, D6S228E
Chromosome Location
Chr.6: 69866556 - 70215063 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001858.5 NP_001849.2 47 - 48 3144 153
XM_011535437.2 XP_011533739.1 47 - 48 3411 153
XM_011535438.2 XP_011533740.1 47 - 48 3139 153
XM_017010253.1 XP_016865742.1 46 - 47 3369 153
XM_017010254.1 XP_016865743.1 46 - 47 3357 153
XM_017010255.1 XP_016865744.1 47 - 48 3350 153
XM_017010256.1 XP_016865745.1 46 - 47 3102 153
XM_017010257.1 XP_016865746.1 46 - 47 3285 153
XM_017010258.1 XP_016865747.1 46 - 47 3018 153
GenBank mRNA BC113362.1 - 47 - 48 3112 153
BC113364.1 - 47 - 48 3112 153
D38163.1 - 47 - 48 3144 153
U09279.1 - 15 - 16 816 153

View Full Product Details