Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- MESTIT1
- Entrez Gene ID
-
317751
- Gene Name
- MEST intronic transcript 1, antisense RNA
- Gene Aliases
-
MEST-AS1, MEST-IT, MEST-IT1, NCRNA00040, PEG1-AS
- Chromosome Location
-
Chr.7: 130487057 - 130491172 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_004382.1 |
|
1 - 2 |
337 |
154 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AW161444.1 |
- |
1 - 2 |
119 |
|
Chr. 7 - 130489632 - 130490954 [-] on Build GRCh38 |
154 |
GenBankmRNA |
DB099005.1 |
- |
1 - 2 |
198 |
|
Chr. 7 - 130489574 - 130491033 [-] on Build GRCh38 |
154 |
GenBankmRNA |
DB055510.1 |
- |
1 - 2 |
201 |
|
Chr. 7 - 130489585 - 130491033 [-] on Build GRCh38 |
154 |
GenBankmRNA |
AW163454.1 |
- |
1 - 2 |
119 |
|
Chr. 7 - 130489582 - 130490954 [-] on Build GRCh38 |
154 |
GenBankmRNA |
AF482998.3 |
- |
1 - 2 |
337 |
|
Chr. 7 - 130487057 - 130491172 [-] on Build GRCh38 |
154 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
centrosomal protein 41 |
CEP41 |
JBTS15, TSGA14 |
-47.8kb
|
Sense |
Chr. - 130393771 - 130441210 on Build GRCh38 |
Downstream |
microRNA 335 |
MIR335 |
MIRN335, hsa-mir-335, miRNA335, mir-335 |
5.1kb
|
Anti-sense |
Chr. - 130496111 - 130496204 on Build GRCh38 |
View Full Product Details