Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MUC16
Entrez Gene ID
94025
Gene Name
mucin 16, cell surface associated
Gene Aliases
CA125
Chromosome Location
Chr.19: 8848840 - 9010390 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024690.2 NP_078966.2 5 - 6 36264 69
XM_017027486.1 XP_016882975.1 8 - 9 36214 69
XM_017027487.1 XP_016882976.1 8 - 9 36214 69
XM_017027488.1 XP_016882977.1 8 - 9 36214 69
XM_017027489.1 XP_016882978.1 5 - 6 36060 69
XM_017027490.1 XP_016882979.1 8 - 9 36214 69
XM_017027491.1 XP_016882980.1 8 - 9 36214 69
XM_017027492.1 XP_016882981.1 8 - 9 36214 69
XM_017027493.1 XP_016882982.1 8 - 9 36214 69
XM_017027494.1 XP_016882983.1 8 - 9 36214 69
XM_017027495.1 XP_016882984.1 8 - 9 36214 69
XM_017027496.1 XP_016882985.1 8 - 9 36214 69
XM_017027497.1 XP_016882986.1 8 - 9 36214 69
XM_017027498.1 XP_016882987.1 8 - 9 36214 69
XM_017027499.1 XP_016882988.1 8 - 9 27001 69
XM_017027500.1 XP_016882989.1 7 - 8 26920 69
GenBank mRNA AF361486.3 - - 11178 69
AF414442.2 - - 36270 69

View Full Product Details