The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145913.3 | NP_666018.3 | 13 - 14 | 2015 | 176 |
GenBank mRNA | AF536216.1 | - | 13 - 14 | 1988 | 176 |
AK075263.1 | - | 13 - 14 | 1673 | 176 | |
AK313788.1 | - | 13 - 14 | 1984 | 176 | |
AY081220.1 | - | 13 - 14 | 1989 | 176 | |
BC110492.2 | - | 13 - 14 | 1650 | 176 |