The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145913.3 | NP_666018.3 | 5 - 6 | 1087 | 102 |
XM_017018910.1 | XP_016874399.1 | 5 - 6 | 1123 | 102 | |
GenBank mRNA | AF536216.1 | - | 5 - 6 | 1060 | 102 |
AK075263.1 | - | 5 - 6 | 745 | 102 | |
AK313788.1 | - | 5 - 6 | 1056 | 102 | |
AY081220.1 | - | 5 - 6 | 1061 | 102 | |
BC110492.2 | - | 5 - 6 | 722 | 102 |