The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018557.2 | NP_061027.2 | 52 - 53 | 9368 | 114 |
XM_017004341.1 | XP_016859830.1 | 52 - 53 | 13944 | 114 | |
XM_017004342.1 | XP_016859831.1 | 23 - 24 | 3367 | 114 | |
GenBank mRNA | AF176832.1 | - | 52 - 53 | 9368 | 114 |