Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BRIX1
Entrez Gene ID
55299
Gene Name
BRX1, biogenesis of ribosomes
Gene Aliases
BRIX, BXDC2
Chromosome Location
Chr.5: 34915715 - 34925682 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018321.3 NP_060791.3 4 - 5 413 149
GenBank mRNA AF347667.1 - 4 - 5 389 149
AK289610.1 - 4 - 5 412 149
AK303311.1 - 4 - 5 452 149
AY364244.1 - 4 - 5 389 149
BC001546.1 - 4 - 5 413 149
BC006014.1 - 4 - 5 416 149
BC036741.1 - 4 - 5 763 149

View Full Product Details