Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ORC1
Entrez Gene ID
4998
Gene Name
origin recognition complex subunit 1
Gene Aliases
HSORC1, ORC1L, PARC1
Chromosome Location
Chr.1: 52371408 - 52404471 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190818.1 NP_001177747.1 15 - 16 2532 85
NM_001190819.1 NP_001177748.1 15 - 16 2520 85
NM_004153.3 NP_004144.2 15 - 16 2535 85
XM_011541527.2 XP_011539829.1 15 - 16 2484 85
XM_017001388.1 XP_016856877.1 14 - 15 2427 85
XM_017001389.1 XP_016856878.1 16 - 17 2572 85
GenBank mRNA AK291909.1 - 15 - 16 2510 85
AK303420.1 - 15 - 16 2461 85
BC011539.1 - 15 - 16 2448 85
U40152.1 - 15 - 16 2523 85
U43416.1 - 15 - 16 2515 85

View Full Product Details