The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005236.2 | NP_005227.1 | 8 - 9 | 1823 | 75 |
XM_011522424.2 | XP_011520726.1 | 9 - 10 | 1980 | 75 | |
XM_011522427.1 | XP_011520729.1 | 3 - 4 | 960 | 75 | |
XM_017023043.1 | XP_016878532.1 | 9 - 10 | 1937 | 75 | |
GenBank mRNA | AK289726.1 | - | 8 - 9 | 1822 | 75 |
AK301930.1 | - | 3 - 4 | 1092 | 75 | |
U64315.1 | - | 8 - 9 | 1796 | 75 |