Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
JAG1
Entrez Gene ID
182
Gene Name
jagged 1
Gene Aliases
AGS, AHD, AWS, CD339, HJ1, JAGL1
Chromosome Location
Chr.20: 10637684 - 10674046 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000214.2 NP_000205.1 25 - 26 3713 74
XM_017027707.1 XP_016883196.1 24 - 25 3818 74
GenBank mRNA AF003837.1 - 25 - 26 3656 74
AF028593.1 - 25 - 26 3460 74
AK302554.1 - 23 - 24 3063 74
AK312722.1 - 25 - 26 3282 74
BC098393.1 - 25 - 26 3616 74
BC126205.1 - 26 - 27 3221 74
BC126207.1 - 25 - 26 3449 74
U61276.1 - 25 - 26 3567 74
U73936.1 - 25 - 26 3610 74
U77720.1 - 25 - 26 3159 74
U77914.1 - 8 - 9 883 74

View Full Product Details