The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100426.1 | NP_001093896.1 | 7 - 8 | 946 | 137 |
NM_001100427.1 | NP_001093897.1 | 7 - 8 | 943 | 137 | |
NM_001100428.1 | NP_001093898.1 | 6 - 7 | 799 | 137 | |
NM_001100429.1 | NP_001093899.1 | 6 - 7 | 796 | 137 | |
NM_001100430.1 | NP_001093900.1 | 5 - 6 | 670 | 137 | |
NM_021159.4 | NP_066982.3 | 7 - 8 | 946 | 137 | |
GenBank mRNA | AF215923.1 | - | 6 - 7 | 753 | 137 |
AF237413.1 | - | 6 - 7 | 753 | 137 | |
AK055032.1 | - | 7 - 8 | 866 | 137 | |
AK295156.1 | - | 6 - 7 | 758 | 137 | |
AK297543.1 | - | 4 - 5 | 493 | 137 | |
AK311243.1 | - | 4 - 5 | 481 | 137 | |
AY376900.1 | - | 4 - 5 | 480 | 137 | |
BC001816.2 | - | 7 - 8 | 853 | 137 | |
BC001851.2 | - | 7 - 8 | 853 | 137 | |
BC098269.1 | - | 7 - 8 | 804 | 137 | |
BC098334.1 | - | 6 - 7 | 654 | 137 | |
BC099708.1 | - | 6 - 7 | 654 | 137 | |
BC099845.1 | - | 6 - 7 | 657 | 137 | |
BT006837.1 | - | 6 - 7 | 756 | 137 | |
X63465.1 | - | 5 - 6 | 606 | 137 |