The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152380.2 | NP_689593.2 | 4 - 5 | 694 | 108 |
XM_005271161.3 | XP_005271218.1 | 4 - 5 | 1150 | 108 | |
XM_005271162.1 | XP_005271219.1 | 4 - 5 | 1150 | 108 | |
GenBank mRNA | AK127536.1 | - | 4 - 5 | 694 | 108 |
BC122553.1 | - | 4 - 5 | 512 | 108 |