Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBX15
Entrez Gene ID
6913
Gene Name
T-box 15
Gene Aliases
TBX14
Chromosome Location
Chr.1: 118883043 - 118989556 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152380.2 NP_689593.2 6 - 7 922 118
XM_005271161.3 XP_005271218.1 6 - 7 1378 118
XM_005271162.1 XP_005271219.1 6 - 7 1378 118
GenBank mRNA AK127536.1 - 6 - 7 922 118
BC122553.1 - 6 - 7 740 118

View Full Product Details