Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SH2D5
Entrez Gene ID
400745
Gene Name
SH2 domain containing 5
Gene Aliases
-
Chromosome Location
Chr.1: 20719731 - 20732640 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103160.1 NP_001096630.1 2 - 3 535 126
NM_001103161.1 NP_001096631.1 3 - 4 664 126
XM_011541459.1 XP_011539761.1 3 - 4 1129 126
XM_011541460.1 XP_011539762.1 4 - 5 969 126
XM_011541461.2 XP_011539763.1 3 - 4 242 126
XM_011541462.1 XP_011539764.1 3 - 4 1129 126
GenBank mRNA AK124869.1 - 2 - 3 535 126
AK296431.1 - 3 - 4 233 126
DC382086.1 - 3 - 4 250 126

View Full Product Details