The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001103160.1 | NP_001096630.1 | 4 - 5 | 765 | 91 |
NM_001103161.1 | NP_001096631.1 | 5 - 6 | 894 | 91 | |
XM_011541459.1 | XP_011539761.1 | 5 - 6 | 1359 | 91 | |
XM_011541460.1 | XP_011539762.1 | 6 - 7 | 1199 | 91 | |
XM_011541461.2 | XP_011539763.1 | 5 - 6 | 472 | 91 | |
XM_011541462.1 | XP_011539764.1 | 5 - 6 | 1359 | 91 | |
GenBank mRNA | AK124869.1 | - | 4 - 5 | 765 | 91 |
AK296431.1 | - | 5 - 6 | 463 | 91 | |
DC382086.1 | - | 5 - 6 | 480 | 91 |