The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014058.3 | NP_054777.2 | 3 - 4 | 316 | 137 |
XM_011531896.2 | XP_011530198.1 | 2 - 3 | 166 | 137 | |
GenBank mRNA | AF064819.1 | - | 3 - 4 | 304 | 137 |
AK292787.1 | - | 3 - 4 | 302 | 137 | |
AK313859.1 | - | 3 - 4 | 316 | 137 | |
AY359017.1 | - | 3 - 4 | 283 | 137 | |
BC113412.1 | - | 3 - 4 | 275 | 137 | |
BC113414.1 | - | 3 - 4 | 275 | 137 | |
DB013544.1 | - | 3 - 4 | 316 | 137 |