The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014058.3 | NP_054777.2 | 7 - 8 | 778 | 91 |
XM_011531896.2 | XP_011530198.1 | 6 - 7 | 628 | 91 | |
GenBank mRNA | AF064819.1 | - | 7 - 8 | 766 | 91 |
AK292787.1 | - | 7 - 8 | 764 | 91 | |
AK313859.1 | - | 7 - 8 | 778 | 91 | |
AY359017.1 | - | 7 - 8 | 745 | 91 | |
BC113412.1 | - | 7 - 8 | 737 | 91 | |
BC113414.1 | - | 7 - 8 | 737 | 91 |