The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173814.5 | NP_776175.2 | 5 - 6 | 1003 | 126 |
XM_011521459.1 | XP_011519761.1 | 5 - 6 | 1533 | 126 | |
XM_011521460.1 | XP_011519762.1 | 5 - 6 | 1533 | 126 | |
XM_017022081.1 | XP_016877570.1 | 5 - 6 | 1090 | 126 | |
XM_017022082.1 | XP_016877571.1 | 5 - 6 | 1533 | 126 | |
GenBank mRNA | AY630258.1 | - | 5 - 6 | 863 | 126 |