The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289080.1 | NP_001276009.1 | 21 - 22 | 3369 | 153 |
NM_001289081.1 | NP_001276010.1 | 20 - 21 | 3232 | 153 | |
NM_014461.3 | NP_055276.1 | 21 - 22 | 3378 | 153 | |
XM_005265058.3 | XP_005265115.1 | 22 - 23 | 3501 | 153 | |
XM_011533590.2 | XP_011531892.1 | 21 - 22 | 3025 | 153 | |
XM_011533591.2 | XP_011531893.1 | 20 - 21 | 3340 | 153 | |
XM_017006171.1 | XP_016861660.1 | 26 - 27 | 3928 | 153 | |
XM_017006172.1 | XP_016861661.1 | 21 - 22 | 3503 | 153 | |
XM_017006173.1 | XP_016861662.1 | 20 - 21 | 3009 | 153 | |
XM_017006174.1 | XP_016861663.1 | 20 - 21 | 2992 | 153 | |
XM_017006175.1 | XP_016861664.1 | 18 - 19 | 2666 | 153 | |
XM_017006176.1 | XP_016861665.1 | 19 - 20 | 2810 | 153 | |
GenBank mRNA | AB003592.1 | - | 21 - 22 | 3091 | 153 |
AK294786.1 | - | 20 - 21 | 3232 | 153 | |
AK314220.1 | - | 21 - 22 | 3092 | 153 | |
BC113118.1 | - | 21 - 22 | 3004 | 153 |