The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289080.1 | NP_001276009.1 | 22 - 23 | 3538 | 91 |
NM_001289081.1 | NP_001276010.1 | 21 - 22 | 3401 | 91 | |
NM_014461.3 | NP_055276.1 | 22 - 23 | 3547 | 91 | |
XM_005265058.3 | XP_005265115.1 | 23 - 24 | 3670 | 91 | |
XM_011533590.2 | XP_011531892.1 | 22 - 23 | 3194 | 91 | |
XM_011533591.2 | XP_011531893.1 | 21 - 22 | 3509 | 91 | |
XM_017006171.1 | XP_016861660.1 | 27 - 28 | 4097 | 91 | |
XM_017006172.1 | XP_016861661.1 | 22 - 23 | 3672 | 91 | |
XM_017006173.1 | XP_016861662.1 | 21 - 22 | 3178 | 91 | |
XM_017006174.1 | XP_016861663.1 | 21 - 22 | 3161 | 91 | |
XM_017006175.1 | XP_016861664.1 | 19 - 20 | 2835 | 91 | |
XM_017006176.1 | XP_016861665.1 | 20 - 21 | 2979 | 91 | |
GenBank mRNA | AB003592.1 | - | 22 - 23 | 3260 | 91 |
AK294786.1 | - | 21 - 22 | 3401 | 91 | |
AK314220.1 | - | 22 - 23 | 3261 | 91 | |
BC113118.1 | - | 22 - 23 | 3173 | 91 | |
CN364480.1 | - | 1 - 2 | 61 | 91 |