The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001006616.2 | NP_001006617.2 | 4 - 5 | 685 | 107 |
NM_012171.2 | NP_036303.1 | 4 - 5 | 685 | 107 | |
NM_130465.4 | NP_569732.2 | 4 - 5 | 685 | 107 | |
XM_005265876.3 | XP_005265933.1 | 2 - 3 | 467 | 107 | |
GenBank mRNA | BC010405.2 | - | 4 - 5 | 507 | 107 |
BC067105.1 | - | 4 - 5 | 570 | 107 |