The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005956.3 | NP_005947.3 | 26 - 27 | 3111 | 148 |
GenBank mRNA | BC001014.2 | - | 9 - 10 | 1009 | 148 |
BC009806.2 | - | 26 - 27 | 2777 | 148 | |
BC050420.1 | - | 26 - 27 | 2802 | 148 | |
CA446032.1 | - | 2 - 3 | 148 | ||
J04031.1 | - | 26 - 27 | 2777 | 148 |