The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320845.1 | NP_001307774.1 | 3 - 4 | 459 | 130 |
NM_133491.4 | NP_597998.1 | 3 - 4 | 442 | 130 | |
XM_017024073.1 | XP_016879562.1 | 3 - 4 | 459 | 130 | |
GenBank mRNA | AF348524.1 | - | 3 - 4 | 201 | 130 |
BC011751.2 | - | 3 - 4 | 412 | 130 | |
BC095466.1 | - | 3 - 4 | 397 | 130 |