The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320845.1 | NP_001307774.1 | 5 - 6 | 600 | 72 |
NM_001320846.1 | NP_001307775.1 | 4 - 5 | 498 | 72 | |
NM_001320847.1 | NP_001307776.1 | 4 - 5 | 481 | 72 | |
NM_133491.4 | NP_597998.1 | 5 - 6 | 583 | 72 | |
GenBank mRNA | AF348524.1 | - | 5 - 6 | 342 | 72 |
BC011751.2 | - | 5 - 6 | 553 | 72 | |
BC095466.1 | - | 5 - 6 | 538 | 72 | |
BG748008.1 | - | 4 - 5 | 408 | 72 | |
BP317593.1 | - | 4 - 5 | 499 | 72 | |
HY155008.1 | - | 4 - 5 | 484 | 72 |