Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SETD4
Entrez Gene ID
54093
Gene Name
SET domain containing 4
Gene Aliases
C21orf18, C21orf27
Chromosome Location
Chr.21: 36034541 - 36060818 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017438.4 NP_059134.1 11 - 12 1863 65
NR_040087.2 10 - 11 1774 65
XM_011529636.1 XP_011527938.1 11 - 12 1912 65
XM_011529637.1 XP_011527939.1 11 - 12 1977 65
XM_011529638.2 XP_011527940.1 11 - 12 1885 65
XM_011529639.1 XP_011527941.1 11 - 12 1663 65
XM_011529640.2 XP_011527942.1 11 - 12 1928 65
XM_011529642.1 XP_011527944.1 12 - 13 2006 65
XM_011529643.1 XP_011527945.1 12 - 13 1957 65
XM_011529644.1 XP_011527946.1 12 - 13 1518 65
XM_017028404.1 XP_016883893.1 12 - 13 1562 65
GenBank mRNA AF391112.1 - 10 - 11 1341 65
AK001660.1 - 11 - 12 1430 65
AK126368.1 - 4 - 5 1940 65

View Full Product Details