The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020665.5 | NP_065716.1 | 1 - 2 | 312 | 94 |
XM_017029681.1 | XP_016885170.1 | 1 - 2 | 587 | 94 | |
GenBank mRNA | AF229179.1 | - | 1 - 2 | 79 | 94 |
AK313835.1 | - | 1 - 2 | 94 | ||
AY359060.1 | - | 1 - 2 | 62 | 94 | |
BC014317.1 | - | 1 - 2 | 316 | 94 | |
BC015099.1 | - | 1 - 2 | 144 | 94 | |
BC050606.1 | - | 1 - 2 | 64 | 94 |