Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM27
Entrez Gene ID
57393
Gene Name
transmembrane protein 27
Gene Aliases
NX-17, NX17
Chromosome Location
Chr.X: 15627316 - 15675624 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020665.5 NP_065716.1 5 - 6 764 139
XM_017029680.1 XP_016885169.1 5 - 6 632 139
GenBank mRNA AF229179.1 - 5 - 6 531 139
AK313835.1 - 5 - 6 589 139
AY359060.1 - 5 - 6 514 139
BC014317.1 - 5 - 6 768 139
BC015099.1 - 5 - 6 596 139
BC050606.1 - 5 - 6 516 139

View Full Product Details