The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278064.1 | NP_001264993.1 | 7 - 8 | 2985 | 102 |
XM_011535782.1 | XP_011534084.1 | 8 - 9 | 3135 | 102 | |
XM_017010783.1 | XP_016866272.1 | 8 - 9 | 3144 | 102 | |
XM_017010784.1 | XP_016866273.1 | 8 - 9 | 3180 | 102 | |
XM_017010788.1 | XP_016866277.1 | 6 - 7 | 1952 | 102 | |
GenBank mRNA | BC136280.1 | - | 7 - 8 | 2697 | 102 |
L76627.1 | - | 7 - 8 | 2665 | 102 | |
U31215.1 | - | 7 - 8 | 2900 | 102 |