The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014171.5 | NP_054890.1 | 2 - 3 | 192 | 162 |
GenBank mRNA | AF161488.1 | - | 2 - 3 | 168 | 162 |
AK291298.1 | - | 2 - 3 | 176 | 162 | |
BC006980.1 | - | 2 - 3 | 149 | 162 | |
BC018653.1 | - | 2 - 3 | 156 | 162 | |
BU603377.1 | - | - | 195 | 162 |