The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001202543.1 | NP_001189472.1 | 3 - 4 | 354 | 81 |
NM_001202545.2 | NP_001189474.1 | 3 - 4 | 354 | 81 | |
NM_001202546.2 | NP_001189475.1 | 2 - 3 | 139 | 81 | |
NM_001913.4 | NP_001904.2 | 3 - 4 | 354 | 81 | |
NM_181500.3 | NP_852477.1 | 3 - 4 | 354 | 81 | |
NM_181552.3 | NP_853530.2 | 3 - 4 | 232 | 81 | |
XM_005250150.2 | XP_005250207.1 | 3 - 4 | 753 | 81 | |
XM_005250151.2 | XP_005250208.1 | 3 - 4 | 756 | 81 | |
XM_005250154.3 | XP_005250211.1 | 3 - 4 | 610 | 81 | |
XM_006715854.2 | XP_006715917.1 | 3 - 4 | 753 | 81 | |
XM_006715855.1 | XP_006715918.1 | 3 - 4 | 610 | 81 | |
XM_011515823.2 | XP_011514125.1 | 3 - 4 | 751 | 81 | |
XM_011515825.1 | XP_011514127.1 | 3 - 4 | 319 | 81 | |
XM_017011759.1 | XP_016867248.1 | 3 - 4 | 319 | 81 | |
XM_017011760.1 | XP_016867249.1 | 3 - 4 | 319 | 81 | |
GenBank mRNA | AK122726.1 | - | 3 - 4 | 237 | 81 |
AK222832.1 | - | 3 - 4 | 250 | 81 | |
AK297548.1 | - | 3 - 4 | 240 | 81 | |
AK303151.1 | - | 2 - 3 | 143 | 81 | |
BC025422.2 | - | 3 - 4 | 235 | 81 | |
BC066592.1 | - | 3 - 4 | 247 | 81 | |
L12579.1 | - | 3 - 4 | 246 | 81 | |
M74099.1 | - | 3 - 4 | 237 | 81 |