Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POC5
Entrez Gene ID
134359
Gene Name
POC5 centriolar protein
Gene Aliases
C5orf37
Chromosome Location
Chr.5: 75674198 - 75717488 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099271.1 NP_001092741.1 6 - 7 878 124
NM_152408.2 NP_689621.2 5 - 6 677 124
XM_005248436.1 XP_005248493.1 6 - 7 878 124
XM_011543158.2 XP_011541460.1 6 - 7 878 124
XM_011543159.1 XP_011541461.1 7 - 8 884 124
XM_011543160.1 XP_011541462.1 5 - 6 738 124
XM_017009038.1 XP_016864527.1 5 - 6 798 124
XM_017009039.1 XP_016864528.1 5 - 6 717 124
GenBank mRNA AK093098.1 - 5 - 6 647 124
AK296071.1 - 6 - 7 795 124
BC065750.1 - 6 - 7 824 124
BC101325.1 - 5 - 6 643 124
BC101326.2 - 5 - 6 647 124
BC101327.2 - 5 - 6 647 124
BC101328.2 - 5 - 6 644 125

View Full Product Details