Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PABPC1P2
- Entrez Gene ID
-
728773
- Gene Name
- poly(A) binding protein cytoplasmic 1 pseudogene 2
- Gene Aliases
-
PABP2, PABP4, PABPCP2, PABPCP4
- Chromosome Location
-
Chr.2: 146587057 - 146590990 on Build GRCh38
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
testis expressed 41 (non-protein coding) |
TEX41 |
DKFZp686O1327, LINC00953 |
-1.5mb
|
Sense |
Chr. - 144667967 - 145076729 on Build GRCh38 |
Downstream |
activin A receptor type 2A |
ACVR2A |
ACTRII, ACVR2 |
1.3mb
|
Sense |
Chr. - 147844517 - 147930827 on Build GRCh38 |
View Full Product Details