Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBP1
Entrez Gene ID
7342
Gene Name
upstream binding protein 1 (LBP-1a)
Gene Aliases
LBP-1B, LBP-1a, LBP1A, LBP1B
Chromosome Location
Chr.3: 33388336 - 33440405 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128160.1 NP_001121632.1 2 - 3 792 90
NM_001128161.1 NP_001121633.1 3 - 4 411 90
NM_014517.4 NP_055332.3 2 - 3 792 90
GenBank mRNA AF198487.1 - 2 - 3 436 90
AK293100.1 - 3 - 4 411 90
BC047235.1 - 2 - 3 791 90
DB131031.1 - 3 - 4 384 90

View Full Product Details