Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBP1
Entrez Gene ID
7342
Gene Name
upstream binding protein 1 (LBP-1a)
Gene Aliases
LBP-1B, LBP-1a, LBP1A, LBP1B
Chromosome Location
Chr.3: 33388336 - 33440405 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128160.1 NP_001121632.1 6 - 7 1240 77
NM_001128161.1 NP_001121633.1 7 - 8 859 77
NM_014517.4 NP_055332.3 6 - 7 1240 77
GenBank mRNA AF198487.1 - 6 - 7 884 77
AK023274.1 - 6 - 7 869 77
AK023278.1 - 1 - 2 150 77
AK293100.1 - 7 - 8 859 77
BC047235.1 - 6 - 7 1239 77

View Full Product Details