The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080927.3 | NP_563615.3 | 13 - 14 | 2031 | 89 |
XM_011512419.2 | XP_011510721.1 | 12 - 13 | 1747 | 89 | |
GenBank mRNA | AB073146.1 | - | 13 - 14 | 2031 | 89 |
AF387547.1 | - | 13 - 14 | 1679 | 89 | |
AK302649.1 | - | 13 - 14 | 1511 | 89 | |
BC029658.1 | - | 13 - 14 | 1550 | 89 |