Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PCOLCE
Entrez Gene ID
5118
Gene Name
procollagen C-endopeptidase enhancer
Gene Aliases
PCPE, PCPE-1, PCPE1
Chromosome Location
Chr.7: 100602259 - 100608175 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002593.3 NP_002584.2 8 - 9 1374 68
GenBank mRNA AB008549.1 - 8 - 9 1232 68
BC000574.2 - 8 - 9 1267 68
BC033205.1 - 8 - 9 1212 68
L33799.1 - 8 - 9 1236 68

View Full Product Details