The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256891.1 | NP_001243820.1 | 1 - 2 | 111 | 74 |
NM_003130.3 | NP_003121.1 | 1 - 2 | 111 | 74 | |
GenBank mRNA | AK129770.1 | - | 1 - 2 | 80 | 74 |
AK295225.1 | - | 1 - 2 | 64 | 74 | |
AK310366.1 | - | 1 - 2 | 87 | 74 | |
AL039198.1 | - | 1 - 2 | 106 | 74 | |
BC011025.1 | - | 1 - 2 | 69 | 74 | |
BI524104.1 | - | 1 - 2 | 71 | 74 | |
BI559592.1 | - | 1 - 2 | 105 | 74 | |
BI597920.1 | - | 1 - 2 | 107 | 74 | |
DA268050.1 | - | 1 - 2 | 111 | 74 | |
L12387.1 | - | 1 - 2 | 57 | 74 | |
M32886.1 | - | 1 - 2 | 65 | 74 |